Learn More
Christeyns™ Phago'Surf 2D
Product Code. 17850265
Change view
Click to view available options
Size:
5 L
20 L
Unit Size:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17850265 20 L 20 L
17860265 5 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17850265 Supplier Christeyns™ Supplier No. C60425
  • ONLINE EXCLUSIVE PRICE
    210.00 EUR
    182.88 EUR / 20 L
    Save 27.12 EUR
    12% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
Phago'Surf 2D is a concentrated liquid intended for the cleaning and disinfection of floors, surfaces and materials in high-risk environments.
  • Effective cleaner and broad-spectrum disinfectant (bactericidal, yeasticidal, fungicidal, tuberculocidal and active on viruses)
  • Requires rinsing on surfaces likely to come into direct contact with foodstuffs (in accordance with the decree of September 8, 1999)
  • Biocidal product registered in TP2 TP3 and TP4
  • Fast drying, streak-free
  • No scent
  • 'Food contact' product
Specifications
Product Type Disinfectant Detergent
Color-coded No
Form Concentrated Liquid
pH Neutral
Scent Unscented
Sterility Non-sterile
Quantity 1 Can
Size 20 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • PHAGOSURF 2D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-(3-Aminopropyl)-N-dodecylpropane-1,3-diamine, Didecyldimethylammonium chloride, Ethoxylated Laury