Learn More
Christeyns™ Phago'Derm Pure Soap
Product Code. 16873083
Change view
Click to view available options
Volume (Metric):
500 mL
1 L
Unit Size:
12 L
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16873083 500 mL Pack of 20
17860245 1 L 12 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16873083 Supplier Christeyns™ Supplier No. 65072
  • ONLINE EXCLUSIVE PRICE
    66.00 EUR
    57.66 EUR / Pack of 20
    Save 8.34 EUR
    12% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-01-2027
Phago'Derm Pure is intended for the simple washing of hands and forearms in intensive use.
  • Also suitable for patient washing and preoperative showering
  • Recommended for sensitive and reactive skin
  • Square bottle with mounted pump
Specifications
Product Type Hand Cleanser
Color Colorless
Form Liquid
Quantity 20 Bottles
Volume (Metric) 500 mL
compliance-icons compliance-icons
Product Identifier
  • X20 Phago'Derm Pure mild soap 500ml
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol, 1-Propanaminium, Amines, C12-14