Learn More
Macherey-Nagel™ pH-Fix Test Strips in PT Tubes
Product Code. 10025504 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10025504 0 to 14 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10025504 Supplier Macherey-Nagel™ Supplier No. 92111
  • ONLINE EXCLUSIVE PRICE
    16.15 EUR
    14.32 EUR / Pack of 100
    Save 1.83 EUR
    11% Off
View all promotions
 Restricted Item
In Stock
Provides quick and easy pH value testing at the point of interest. Macherey Nagel™ pH-Fix Test Strips in PT Tubes don't need any calibration, set-up or maintenance.
Provides quick and easy pH value testing at the point of interest. Macherey Nagel™ pH-Fix Test Strips in PT Tubes don't need any calibration, set-up or maintenance.
  • Long plastic handle protects against toxic, dangerous or aggressive substances
  • Features up to 4 different indicator pads that cover the whole pH range
  • No bleeding due to color bounded indicator dyes
  • Tube features simple one-hand operation, prevents against jamming, and is virtually unbreakable
Specifications
pH Range 0 to 14
Quantity 100 Pack
Type pH Test Strip