Learn More
Fisherbrand™ pH Microelectrodes
Product Code. 11747088 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11747088 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11747088
  • ONLINE EXCLUSIVE PRICE
    260.00 EUR
    159.15 EUR / Pack of 1
    Save 100.85 EUR
    38% Off
View all promotions
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
Unique pH microelectrode that is ideal for measuring very small sample volumes such as sweat, saliva, and others in capillary tubes, microtiter plates, test tubes, and other easily accessible containers.
  • Small diameter pH electrode
  • For measurement on very small sample volumes: sweat, saliva ...
  • For measuring in easily accessible containers: capillary tube, microtiter plate, test tube 
  • Glass body with S7 head 
Specifications
Description Glass pH Microelectrodes with BNC connector
Connector Types BNC
pH Range 14
Length (Metric) 18 mm