Learn More
Fisherbrand™ pH Indicator Paper Roll
Product Code. 15940872 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
4 to 7
6.4 to 8
8 to 10
Unit Size:
4 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
15940872 4 to 7 4 units
15950872 6.4 to 8 4 units
15960872 8 to 10 4 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15940872
  • ONLINE EXCLUSIVE PRICE
    42.80 EUR
    20.68 EUR / Pack of 4
    Save 22.12 EUR
    51% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
Ready to use universal indicator papers are supplied in rotary lid boxes to ensure protection against external influences.
  • The reaction colour of the paper is compared with the enclosed colour scale and the pH is read off
  • Allows very simple pH measurement
  • Proven standard for many applications
Specifications
pH Range 4 to 7
For Use With (Application) pH testing
Graduations 4.0, 4,3, 4,6, 4.9, 5.2, 5.5, 5.8, 6.1, 6.4, 6.7, 7.0
Type pH Indicator Paper