Learn More
pH Electrode Storage Solution, Reagecon™
BIOPHARMA
Product Code. 11533182 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11533182 500 mL 500mL
12890327 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11533182 Supplier Reagecon Supplier No. ESS5
 Restricted Item
In Stock
Add to basket
Add to basket
Designed to maintain proper functioning and accurate readings of pH and reference electrodes. pH Electrode Storage Solution, Reagecon™ is a 3M KCl solution that helps maintain proper working conditions after cleaning or when the electrode has not been in use for an extended time period.
The response, accuracy and lifespan of pH electrodes can be substantially improved through correct handling, cleaning and storage. Designed for routine equipment maintenance, these solutions are safe to use with any sensitive electrode. Electrode storage solutions are made from potassium chloride, free from silver ion, and are suitable for all Blueline electrodes.
Specifications
Composition Potassium Chloride solution, without Silver ion
Concentration 3 mol/L KCl
For Use With (Equipment) pH and Reference Electrodes, All Blueline Electrodes
Filling Solution Type Electrode Storage
Quantity 500 mL