Learn More
Fisherbrand™ pH Electrode
Product Code. 11705628
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11705628 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 11705628 Supplier Fisherbrand™ Supplier No. 5732526B010BB
  • ONLINE EXCLUSIVE PRICE
    44.25 EUR
    34.30 EUR / Pack of 1
    Save 9.95 EUR
    22% Off
 Restricted Item
Estimated Shipment: 12-02-2027
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Offered in a variety of styles including different reference solutions and reference chambers. Fisherbrand™ pH Electrode feature a refillable outer reference reservoir that can be filled with any electrolyte compatible with the sample, including those fills recommended for nonaqueous measurements.
  • Double junction reference is recommended for samples containing organic compounds, proteins, heavy metals or compounds that interact with silver (e.g. sulfide, bromide, iodide)
  • All electrodes are available with a detachable lead or with a cable/connector attached
  • Designed for all existing Brinkmann, Metrohm and Schott titration systems
  • The electrode body is constructed with a glass ground joint that will fit their titration vessels
  • Ionics and Tytronics titrators can be fitted with this same electrode without the ground joint
  • Either design can be ordered depending on the titration unit
Specifications
Color Blue
Temperature (Metric) Range 0°C to 80°C