Learn More
Hanna Instruments™ pH Edge™ Electrode
Product Code. 15188239
Change view
Click to view available options
Connector Types:
3.5 mm Jack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
15188239 3.5 mm Jack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15188239 Supplier Hanna Instruments™ Supplier No. HI12300
  • ONLINE EXCLUSIVE PRICE
    289.19 EUR
    230.00 EUR / Pack of 1
    Save 59.19 EUR
    20% Off
Estimated Shipment: 10-12-2026
Specifications
Connector Types 3.5 mm Jack
Junction Ceramic Single, 15 to 20 μL/h
Material PEI
pH Range 0 to 13
Temperature (Metric) Operating -5°C to +70°C
Length (Metric) Cable 1 m
Diameter (Metric) Cap 7.5 mm