missing translation for 'onlineSavingsMsg'
Learn More

PH-064, MedChemExpress

Product Code. 16486717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486717 5 mg 5mg
16465006 100 mg 100mg
16464008 10 mg 10mg
16410367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486717 Supplier Medchem Express Supplier No. HY10499

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PH-064 (BIM-46187) is an inhibitor of heterotrimeric G-protein complex.

PH-064 (BIM-46187) is an inhibitor of heterotrimeric G-protein complex.

Specifications

Chemical Name or Material PH-064
CAS 892546-37-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C44H58N8O2S2
Quantity 5 mg
Synonym BIM-46187
Solubility Information DMSO : 100 mg/mL (125.77 mM; Need ultrasonic)
SMILES O=C([C@H](CSSC[C@@H](C(N1CCN2C([C@@H]1CC3CCCCC3)=NC(C4=CC=CC=C4)=C2)=O)N)N)N([C@H]5CC6CCCCC6)CCN(C5=N7)C=C7C8=CC=CC=C8
Molecular Weight (g/mol) 795.11
Formula Weight 795.11
Percent Purity 96.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.