Learn More
PFAS Mixture Food Testing EU 2022/1431 10 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Product Code. 30328350
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328350 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328350 Supplier Dr. Ehrenstorfer Supplier No. DREA30000074MW
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 763051-92-9, 919005-14-4, 756426-58-1, 34455-29-3, 80475-32-7, 13252-13-6
Quantity 1 mL
Certifications/Compliance ISO 17034