Learn More
BrandTech™ VITLAB™ PFA Round Bottom Flasks
Product Code. 10384311
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10384311 500 mL Each
10777991 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10384311 Supplier BrandTech™ Supplier No. 107997
  • ONLINE EXCLUSIVE PRICE
    166.00 EUR
    135.18 EUR / Pack of 1
    Save 30.82 EUR
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-01-2027
Take advantage of PFA, which is especially suitable for use in trace analysis, with Vitlab™ PFA Round Bottom Flasks. They have high thermal stability, from -200 °C to +260 °C, and are autoclavable.
  • Transparent
  • Neck with NS 29/32
  • Suitable as safety flask
  • For use with rotary evaporators (operation at room temperature) to collect distilled-off liquid.
  • High thermal stability and chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Material PFA
For Use With (Application) Suitable for use in trace analysis
No. per Pack 1