Learn More
Bohlender™ PFA Jar with Screw Cap
Product Code. 10480801
Change view
Click to view available options
Capacity (Metric):
7 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10480801 7 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10480801 Supplier Bohlender™ Supplier No. A 13001
  • ONLINE EXCLUSIVE PRICE
    61.50 EUR
    50.79 EUR / Pack of 1
    Save 10.71 EUR
    17% Off
Estimated Shipment: 04-01-2027
Specifications
Quantity 1Pk.
Depth (Metric) 32 mm
Capacity (Metric) 7 mL
Color Translucent
Product Type Jars
Material PFA
Height (Metric) 37 mm