missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 06281355, >98%
Description
Selective myeloperoxidase (MPO) inhibitor (IC50 = 1.5 μM in LPS-stimulated human whole blood). Selective for MPO over a panel of 50 receptors, enzymes, ion channels and transporters, including thyroid peroxidase. Inhibits MPO activity in peritoneal lavage and plasma in a murine peritonitis model. Also reduces lung edema in a murine pulmonary immune complex vasculitis model. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Solubility | Soluble to 20mM in DMSO with gentle warming |
| Formula Weight | 321.35 |
| Purity | >98% |
| Molecular Weight (g/mol) | 321.351 |
| Chemical Name or Material | 6-(2,5-Dimethoxyphenyl)-3,4-dihydro-4-oxo-2-thioxo-1(2H)-pyrimidineacetamide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?