missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 06281355, >98%

Product Code. 30000984
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000984 50 mg 50mg
30000902 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30000984 Supplier Tocris Bioscience™ Supplier No. 6004/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective myeloperoxidase (MPO) inhibitor

Selective myeloperoxidase (MPO) inhibitor (IC50 = 1.5 μM in LPS-stimulated human whole blood). Selective for MPO over a panel of 50 receptors, enzymes, ion channels and transporters, including thyroid peroxidase. Inhibits MPO activity in peritoneal lavage and plasma in a murine peritonitis model. Also reduces lung edema in a murine pulmonary immune complex vasculitis model. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1435467-38-1
Molecular Formula C14H15N3O4S

Specifications

Quantity 50 mg
Solubility Soluble to 20mM in DMSO with gentle warming
Formula Weight 321.35
Purity >98%
Molecular Weight (g/mol) 321.351
Chemical Name or Material 6-(2,5-Dimethoxyphenyl)-3,4-dihydro-4-oxo-2-thioxo-1(2H)-pyrimidineacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.