Learn More
PF-05180999, MedChemExpress
Product Code. 16494428
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16494428
Brand: Medchem Express HY111371
MedChemExpress PF-05180999 (PF-999) is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM.
PF-05180999 (PF-999) is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM.
Specifications
Chemical Name or Material PF-05180999
CAS 1394033-54-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H17F3N8
Quantity 10 mg
Synonym PF-999
Solubility Information DMSO : 50 mg/mL (120.66 mM; Need ultrasonic)
SMILES CC1=NC(C2=C(C3=CC=C(C(F)(F)F)C=N3)N(C)N=C2)=C4N1N=CN=C4N5CCC5
Molecular Weight (g/mol) 414.39
Formula Weight 414.39
Percent Purity 98.93%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation