missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 05180999
GREENER_CHOICE

Product Code. p-7116137
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18763999 25 mg 25mg
18716278 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18763999 Supplier Tocris Bioscience™ Supplier No. 6405/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PDE2A inhibitor

PF 05180999 is a potent and selective PDE2A inhibitor (IC50 = 1 nM). Exhibits 2000-fold selectivity for PDE2A over PDE10A. Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(1-Azetidinyl)-7-methyl-5-[1-methyl-5-[5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazol-4-yl]-imidazo[5,1-f][1,2,4]triazine
CAS 1394033-54-5
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C19H17F3N8
Formula Weight Observed MW: 414.39
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.