Learn More
Petroleum, Supelco™
Product Code. 15875390
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15875390 1 L 1 L
15837770 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15875390 Supplier Supelco Supplier No. 1097181000
 Restricted Item
Estimated Shipment: 14-12-2026
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
For analysis EMSURE™
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64742-48-9
Specifications
Melting Point <-50°C
Density 0.749 to 0.761 g/cm3 at 25°C
Quantity 1 L
MDL Number MFCD32062494
Synonym Petroleum, Petroleum
Grade Research
Chemical Name or Material Petroleum