missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture 651 600 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30328529
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328529 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328529 Supplier Dr. Ehrenstorfer Supplier No. DRES50000651AL
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetamiprid∣Azoxystrobin∣Bifenazate∣Boscalid∣Chlorfenapyr∣Etoxazole∣Fludioxonil∣Kresoxim-methyl∣Metalaxyl∣MGK 264∣Piperonyl butoxide∣Spiromesifen∣Spiroxamine∣Trifloxystrobin
CAS 135410-20-7, 131860-33-8, 149877-41-8, 188425-85-6, 122453-73-0, 153233-91-1
Analyte or Component Names Acetamiprid∣Azoxystrobin∣Bifenazate∣Boscalid∣Chlorfenapyr∣Etoxazole∣Fludioxonil∣Kresoxim-methyl∣Metalaxyl∣MGK 264∣Piperonyl butoxide∣Spiromesifen∣Spiroxamine∣Trifloxystrobin
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Acetamiprid 600 μg/mL∣Azoxystrobin 600 μg/mL∣Bifenazate 600 μg/mL∣Boscalid 600 μg/mL∣Chlorfenapyr 600 μg/mL∣Etoxazole 600 μg/mL∣Fludioxonil 600 μg/mL∣Kresoxim-methyl 600 μg/mL∣Metalaxyl 600 μg/mL∣MGK 264 600 μg/mL∣Piperonyl butoxide 600 μg/mL∣Spiromesifen 600 μg/mL∣Spiroxamine 600 μg/mL∣Trifloxystrobin 600 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.