Learn More
- Home
- Products
- Facility Safety and Maintenance
- Electrical and Lighting Maintenance
- Brady™ PermaSleeve™ Commercial Grade Heat Shrink Wire and Cable Labels for M6/M7 Printers
Brady™ PermaSleeve™ Commercial Grade Heat Shrink Wire and Cable Labels for M6/M7 Printers
Product Code. 17868904
Change view
Click to view available options
Dimensions (L x W):
0.182 x 1 in.
0.182 x 1.75 in.
0.235 x 1 in.
0.235 x 1.75 in.
0.335 x 1 in.
0.335 x 1.75 in.
0.439 x 1 in.
0.439 x 1.75 in.
0.645 x 1 in.
0.645 x 1.75 in.
0.851 x 1 in.
Wire Gauge:
12 to 4 AWG
16 to 8 AWG
20 to 10 AWG
22 to 16 AWG
28 to 20 AWG
8 to 1 AWG
Color:
White
Yellow
Height (Metric):
4.62 mm
5.97 mm
8.51 mm
11.15 mm
16.38 mm
21.62 mm
Unit Size:
Pack of 1000
Pack of 2500
18 product options available for selection
| Product Code. | Dimensions (L x W) | Wire Gauge | Color | Height (Metric) | unitSize |
|---|---|---|---|---|---|
| 17868904 | 0.182 x 1 in. | 28 to 20 AWG | White | 4.62 mm | Pack of 2500 |
| 17839044 | 0.182 x 1.75 in. | 28 to 20 AWG | White | 4.62 mm | Pack of 2500 |
| 17828324 | 0.235 x 1 in. | 22 to 16 AWG | White | 5.97 mm | Pack of 1000 |
| 17818354 | 0.235 x 1.75 in. | 22 to 16 AWG | White | 5.97 mm | Pack of 1000 |
| 17898454 | 0.335 x 1 in. | 20 to 10 AWG | White | 8.51 mm | Pack of 1000 |
| 17888254 | 0.335 x 1.75 in. | 20 to 10 AWG | White | 8.51 mm | Pack of 1000 |
| 17808734 | 0.439 x 1 in. | 16 to 8 AWG | White | 11.15 mm | Pack of 1000 |
| 17868894 | 0.439 x 1.75 in. | 16 to 8 AWG | White | 11.15 mm | Pack of 1000 |
| 17869094 | 0.645 x 1 in. | 12 to 4 AWG | White | 16.38 mm | Pack of 1000 |
| 17838994 | 0.645 x 1.75 in. | 12 to 4 AWG | White | 16.38 mm | Pack of 1000 |
| 17808834 | 0.851 x 1 in. | 8 to 1 AWG | White | 21.62 mm | Pack of 1000 |
| 17868304 | 0.235 x 1 in. | 22 to 16 AWG | Yellow | 5.97 mm | Pack of 1000 |
| 17868144 | 0.235 x 1.75 in. | 22 to 16 AWG | Yellow | 5.97 mm | Pack of 1000 |
| 17898164 | 0.335 x 1 in. | 20 to 10 AWG | Yellow | 8.51 mm | Pack of 1000 |
| 17828314 | 0.335 x 1.75 in. | 20 to 10 AWG | Yellow | 8.51 mm | Pack of 1000 |
| 17859194 | 0.439 x 1 in. | 16 to 8 AWG | Yellow | 11.15 mm | Pack of 1000 |
| 17888974 | 0.439 x 1.75 in. | 16 to 8 AWG | Yellow | 11.15 mm | Pack of 1000 |
| 17899134 | 0.645 x 1.75 in. | 12 to 4 AWG | Yellow | 16.38 mm | Pack of 1000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Ideal for electrical contractors, maintenance crews, industrial manufacturers and more
- Heat-shrink sleeves provide permanent identification without any extra steps
- Legible legends in temperatures ranging from -55°C to 135°C (-67°F to 275°F)
- UL224 recognized
Specifications
Specifications
| Certifications/Compliance | UL 224 |
| Adhesive Type | Non-adhesive |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Polyolefin |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Electrical Labeling, Wire and Cable Labeling |
| For Use With (Equipment) | M610, M710, M611, BMP61, BMP71 |
| Quantity | 2500 Labels |
| Shape | Rectangle |
| Temperature Range (English) | -67°F to +275°F |
| Temperature Range (Metric) | -55°C to +135°C |
| Wire Gauge | 28 to 20 AWG |
| Dimensions (L x W) | 0.182 x 1 in. |
| Length (English) | 0.182 in. |
| Width (English) | 1 in. |
| Width (Metric) | 25.4 mm |
| Height (Metric) | 4.62 mm |
| Diameter (English) | 0.094 in. |
| Diameter (Metric) | 2.39 mm |
| Alternate Part Number | BM-094-1-312 |
| Show More Show Less |