Learn More
Microspec™ Permanent Marker Pens
Product Code. 16624852
Change view
Click to view available options
Description:
Round Point
Sharp Point
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
16624852 Sharp Point Pack of 10
16634852 Round Point Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16624852 Supplier Microspec™ Supplier No. Z1
Estimated Shipment: 07-12-2026
Available with sharp point or round point
  • 10 markers
  • Permanent
  • Available with sharp point or round point
  • Microspec logo
Specifications
Color White
Description Sharp Point
For Use With (Application) Labeling
Quantity 10