missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, Supelco™

Product Code. 15836860
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15836860 100 g 100g
15826860 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15836860 Supplier Supelco Supplier No. 1005240100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9

Specifications

Melting Point 122°C
Quantity 100 g
MDL Number MFCD00011344
Synonym Periodic acid, Periodic acid
Molecular Weight (g/mol) 227.94
Grade Ungraded
Chemical Name or Material Periodic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.