Learn More
Labcon™ PerformR™ 12x75mm Round Bottom Flow Cytometry Tubes
Product Code. 30280332 Shop All Labcon Products
Change view
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30280332 Blue Pack of 1000
30280329 Green Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30280332 Supplier Labcon™ Supplier No. 33503680009
  • ONLINE EXCLUSIVE PRICE
    48.40 EUR
    30.54 EUR / Pack of 1000
    Save 17.86 EUR
    36% Off
Estimated Shipment: 04-01-2027
Polypropylene tubes for cell sorting and general flow cytometry use
  • Flow Cytometry Tubes
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 3,000 x G
  • Freezable to -80C
  • Autoclavable
  • Capacity: 5 mL
  • Seal with plug or dual position caps sold separately
  • 12 mm W. 75 mm H.
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015, FDA CFR 21 GMP
Color Blue
Product Type Flow Cytometer Accessory