Description
- Flow Cytometry Tubes
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polypropylene USP Class VI resin
- Centrifuging to 3,000 x G
- Freezable to -80C
- Autoclavable
- Capacity: 5 mL
- Seal with plug or dual position caps sold separately
- 12 mm W. 75 mm H.
- Made in USA
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015, FDA CFR 21 GMP |
| Color | Green |
| Product Type | Flow Cytometer Accessory |