Learn More
Liebherr™ Performance Upright Static Freezer With Shelves, for Laboratory Use
Product Code. 17992431 Shop All Liebherr Appliances Products
Change view
Click to view available options
Capacity (Metric):
316 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17992431 316 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 17992431 Supplier Liebherr™ Supplier No. SFFSG 4001 740
  • ONLINE EXCLUSIVE PRICE
    3175.22 EUR
    2906.61 EUR / Pack of 1
    Save 268.61 EUR
    8% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Secure, accurate and ultra energy-efficient freezer storage. Designed for lab applications with Liebherr build-quality you can rely on.
  • Temperature setting: -9°C to -30°C – enables flexible storage of temperature-sensitive items
  • Reliable temperature stability of ±5°C (to IEC 60068-3)
  • SmartFrost – makes defrosting less frequent
  • Noise level: 49 dB – ensures quiet operation in the laboratory
  • Mechanical door lock – prevents unauthorised access
  • Connectivity retrofittable – enables integration with digital monitoring systems
  • Natural refrigerant R 600a – environmentally friendly and energy-efficient
  • Antimicrobial handle – reduces bacteria by 99.99% without additional cleaning effort
  • Max. temperature recording – automatically logs temperature deviations for a quick overview
  • Comprehensive alarm systems – immediate alert in case of malfunction preserves sample quality
  • Ø 10 mm access port for external sensors – easy retrofitting for additional temperature control
  • Self-closing door – closes automatically below 90° and keeps appliance temperature stable
  • 5-year extended warranty option available in select countries
Specifications
Type Laboratory Freezer
Capacity (English) 11.16 cu. ft.
Capacity (Metric) 316 L
Temperature Range -9°C to -30°C
Temperature Resolution 0.1°C
Temperature Sensor NTC
Temperature (Setpoint) -20°C
Temperature Stability ±5 K
Door Style Reversible, Self-closing, Fitted Lock
No. of Doors 1
Door Material Steel
Hinge Side Right
Cabinet Material Scratch-resistant Painted Steel
Chamber Material Polystyrene
Exterior Finish White Powder-coated
Shelves 5
Drawers 6
No. of Drawers 6
Certifications/Compliance ISO 9001, ISO 14001, CE
Style Freestanding
Display Touch Control/Digital Display in Door
Casters Optional (Dia. 80 mm, 100 mm)
Color White
Condenser Type Low-maintenance Rear-wall Wire-on-tube
Control Type Touch Control/Digital Display in Door
For Use With (Application) Storage of Temperature-sensitive Products
Ports One 10 mm Port
Warranty 2 Years Parts and Labor, 5 Year Optional Extended Warranty
Defrost Manual
Lighting None
Dimensions (DxWxH) Exterior 65.4 x 59.7 x 188.4 cm
Monitoring Options Alarms, Remote Alarm Contacts
Access Security Mechanical Door Lock
On-Board Datalogging No
Connectivity WiFi/LAN Module Optional
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Frequency 50 Hz
Phase Single Phase
Wattage 220 W
Energy Usage 385 kWh/a
Compressor CFC-free
Refrigerant R290
Shipping Weight (Metric) 88.7 kg
Weight (Metric) 84.3 kg