Learn More
Heidolph™ Perforated Platform 2000
Product Code. 10613211 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed Range:
0 to 1100 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10613211 0 to 1100 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10613211 Supplier Heidolph™ Supplier No. 5495900000
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 23-11-2026
For use with Unimax 2010, Promax 2020 and Polymax 2040
Universal perforated platform allows for individual arrangement of vessels
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 0 to 1100 rpm
For Use With (Equipment) Unimax 2010, Promax 2020 and Polymax 2040