missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctylmethanol, TRC

Product Code. 30428068
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428068 500 mg
30411742 50 mg
30414127 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428068 Supplier Toronto Research Chemicals Supplier No. TRCP999073500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-56-3
Molecular Formula C9H3OF17
Molecular Weight (g/mol) 450.09
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluoro-1-nonanol, (Heptadecafluorooctyl)methanol, 1,1-Dihydroperfluoro-1-nonanol, 1,1-Dihydroperfluoronon-1-yl Alcohol, 1H,1H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluorononanol
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H3F17O/c10-2(11,1-27)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)26/h27H,1H2
Formula Weight 449.9912
Percent Purity >95%
Chemical Name or Material Perfluorooctylmethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.