Learn More
Perfluorooctylethyl Methacrylate, TRC
Product Code. 17960495
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960495 1 g 1g
17957799 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17960495 Supplier Toronto Research Chemicals Supplier No. TRCP3363081G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1996-88-9
Molecular Formula C14 H9 F17 O2
Molecular Weight (g/mol) 532.191
Synonym Perfluorooctylethyl Methacrylate, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, methacrylate (8CI), Methacrylic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester (7CI, 8CI), 1,1,2,2-Tetrahydroperfluorodecyl methacrylate, 1H,1H,2H,2H-Heptadecafluorodecyl methacrylate, 1H,1H,2H,2H-Perfluorodecyl methacrylate, 2-(Perfluorooctyl)ethyl methacryate, 2-(Perfluorooctyl)ethyl methacrylate, 2-(Perfluorooctyl)ethyl methacylate, 2-Heptadecafluorooctylethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl 2-methylprop-2enoate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl methacrylate, Cheminox FAMAC, Cheminox FAMAC 8, F 17MA, FAMAC, Fluowet 812MA, Fluowet MAE 600, Fluowet MAE 800, FMA 8, Light Ester FM 108, M 1820, M 1820 (ester), MAE 600, MAE 800, MAF 17, Perfluorooctylethyl methacrylate, Viscoat 17FM, 8:2 FTMAC, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, methacrylate (8CI), Methacrylic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester (7CI, 8CI), 1,1,2,2-Tetrahydroperfluorodecyl methacrylate, 1H,1H,2H,2H-Heptadecafluorodecyl methacrylate, 1H,1H,2H,2H-Perfluorodecyl methacrylate, 2-(Perfluorooctyl)ethyl methacryate, 2-(Perfluorooctyl)ethyl methacrylate, 2-(Perfluorooctyl)ethyl methacylate, 2-Heptadecafluorooctylethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl 2-methylprop-2enoate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl methacrylate, Cheminox FAMAC, Cheminox FAMAC 8, F 17MA, FAMAC, Fluowet 812MA, Fluowet MAE 600, Fluowet MAE 800, FMA 8, Light Ester FM 108, M 1820, M 1820 (ester), MAE 600, MAE 800, MAF 17, Perfluorooctylethyl methacrylate, Viscoat 17FM
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H9F17O2/c1-5(2)6(32)33-4-3-7(15,16)8(17,18)9(19,20)10(21,22)11(23,24)12(25,26)13(27,28)14(29,30)31/h1,3-4H2,2H3
Formula Weight 532.0331
Percent Purity >95%
Chemical Name or Material Perfluorooctylethyl Methacrylate
Grade GC