missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctylethyl Methacrylate, TRC

Product Code. 17957799
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17957799 5 g 5g
17960495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17957799 Supplier Toronto Research Chemicals Supplier No. TRCP3363085G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1996-88-9
Molecular Formula C14 H9 F17 O2
Molecular Weight (g/mol) 532.191
Synonym Perfluorooctylethyl Methacrylate, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, methacrylate (8CI), Methacrylic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester (7CI, 8CI), 1,1,2,2-Tetrahydroperfluorodecyl methacrylate, 1H,1H,2H,2H-Heptadecafluorodecyl methacrylate, 1H,1H,2H,2H-Perfluorodecyl methacrylate, 2-(Perfluorooctyl)ethyl methacryate, 2-(Perfluorooctyl)ethyl methacrylate, 2-(Perfluorooctyl)ethyl methacylate, 2-Heptadecafluorooctylethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl 2-methylprop-2enoate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl methacrylate, Cheminox FAMAC, Cheminox FAMAC 8, F 17MA, FAMAC, Fluowet 812MA, Fluowet MAE 600, Fluowet MAE 800, FMA 8, Light Ester FM 108, M 1820, M 1820 (ester), MAE 600, MAE 800, MAF 17, Perfluorooctylethyl methacrylate, Viscoat 17FM, 8:2 FTMAC, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, methacrylate (8CI), Methacrylic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester (7CI, 8CI), 1,1,2,2-Tetrahydroperfluorodecyl methacrylate, 1H,1H,2H,2H-Heptadecafluorodecyl methacrylate, 1H,1H,2H,2H-Perfluorodecyl methacrylate, 2-(Perfluorooctyl)ethyl methacryate, 2-(Perfluorooctyl)ethyl methacrylate, 2-(Perfluorooctyl)ethyl methacylate, 2-Heptadecafluorooctylethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl 2-methylprop-2enoate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl methacrylate, Cheminox FAMAC, Cheminox FAMAC 8, F 17MA, FAMAC, Fluowet 812MA, Fluowet MAE 600, Fluowet MAE 800, FMA 8, Light Ester FM 108, M 1820, M 1820 (ester), MAE 600, MAE 800, MAF 17, Perfluorooctylethyl methacrylate, Viscoat 17FM
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H9F17O2/c1-5(2)6(32)33-4-3-7(15,16)8(17,18)9(19,20)10(21,22)11(23,24)12(25,26)13(27,28)14(29,30)31/h1,3-4H2,2H3
Formula Weight 532.0331
Percent Purity >95%
Chemical Name or Material Perfluorooctylethyl Methacrylate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.