Learn More
Perfluorooctanesulfonyl Chloride, TRC
Product Code. 30410313
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410313 50 mg
30420669 500 mg
30413712 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30410313 Supplier Toronto Research Chemicals Supplier No. TRCP99936050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 423-60-9
Molecular Formula C8ClF17O2S
Molecular Weight (g/mol) 518.58
Synonym 1-Octanesulfonyl chloride, heptadecafluoro- (6CI,7CI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-1-octanesulfonyl chloride, Heptadecafluoro-1-octanesulfonyl chloride, NSC 292152, Perfluoro-1-octanesulfonyl chloride, Perfluorooctylsulfonyl chloride
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonyl chloride
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)Cl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C8ClF17O2S/c9-29(27,28)8(25,26)6(20,21)4(16,17)2(12,13)1(10,11)3(14,15)5(18,19)7(22,23)24
Formula Weight 517.9036
Chemical Name or Material Perfluorooctanesulfonyl Chloride