missing translation for 'onlineSavingsMsg'
Learn More

Perfluorobutylethyl Methacrylate, TRC

Product Code. 30012124
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012124 1 g 1g
30003315 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012124 Supplier Toronto Research Chemicals Supplier No. TRCP2863481G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1799-84-4
Molecular Formula C10 H9 F9 O2
Molecular Weight (g/mol) 332.17
Synonym 2-Perfluorobutylethyl Methacrylate, 1H,1H,2H,2H-Pefluorohexyl Methacrylate, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, FAMAC 4
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexyl methacrylate
SMILES O=C(OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(=C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H9F9O2/c1-5(2)6(20)21-4-3-7(11,12)8(13,14)9(15,16)10(17,18)19/h1,3-4H2,2H3
Formula Weight 332.05
Percent Purity >95%
Chemical Name or Material Perfluorobutylethyl Methacrylate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.