missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanoic-3,3-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Pentanoic-3,3-D2 Acid |
| CAS | 83741-74-6 |
| Molecular Formula | C5 D2 H8 O2 |
| Quantity | 0.5 g |
| Synonym | Pentanoic-3,3-d2 acid (9 CI, ACI), β-Dideuteropentanoic acid, Pentanoic acid-3,3-d2, Valeric acid-d2, Valeric-β,β-d2 acid, Pentanoic-3,3-D2 acid |
| InChI Formula | InChI=1 S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i3D2 |
| SMILES | [2 H]C([2 H])(CC)CC(=O)O |
| IUPAC Name | 3,3-dideuteriopentanoic acid |
| Molecular Weight (g/mol) | 104.144 |
| Formula Weight | 104.081 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction