Learn More
Pentane, ≥99.0%, Solstice™
Product Code. 15697040
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15697040 1 L Poly Bottle 1 L
15607050 2.5 L Poly Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15697040 Supplier Honeywell-Chromasolv Supplier No. 768691L
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CHROMASOLV™, for pesticide residue analysis,≥99.0%
  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
Specifications
pH 5.7 (at 22°C)
Melting Point 750°C
Density 0.626g/mL at 25°C
Quantity 1 L
Explosion Limit 8.3%
Ignition Point 500°C
Packaging Poly Bottle
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Boiling Point 1393°C (1013 hPa)
Solubility Information 946g/L at 25°C
Formula Weight 72.15g/mol
Percent Purity ≥99.0%
Chemical Name or Material Pentane