missing translation for 'onlineSavingsMsg'
Learn More

Pentabromobenzene, TRC

Product Code. 17994605
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994605 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17994605 Supplier Toronto Research Chemicals Supplier No. TRCP220250100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 608-90-2
Molecular Formula C6 H Br5
Quantity 100 mg
Synonym Pentabromobenzene, Benzene, 1,2,3,4,5-pentabromo, Benzene, pentabromo- (7CI,8CI,9CI), 1,2,3,4,5-Pentabromobenzene, Nonnen SMC 65, Pentabromobenzene
InChI Formula InChI=1S/C6HBr5/c7-2-1-3(8)5(10)6(11)4(2)9/h1H
SMILES Brc1cc(Br)c(Br)c(Br)c1Br
IUPAC Name 1,2,3,4,5-pentabromobenzene
Molecular Weight (g/mol) 472.59
Formula Weight 467.5995
Chemical Name or Material Pentabromobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.