Learn More
Penciclovir-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Penciclovir-d4 |
| CAS | 1020719-72-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 2H4 H11 N5 O3 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C10H15N5O3/c11-10-13-8-7(9(18)14-10)12-5-15(8)2-1-6(3-16)4-17/h5-6,16-17H,1-4H2,(H3,11,13,14,18)/i1D2,2D2 |
| SMILES | [2H]C([2H])(C(CO)CO)C([2H])([2H])n1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | 2-amino-9-[1,1,2,2-tetradeuterio-4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one |
| Molecular Weight (g/mol) | 257.28 |
| Formula Weight | 257.1426 |
| Show More |