missing translation for 'onlineSavingsMsg'
Learn More

Pelitinib, MedChemExpress

Product Code. 16498038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498038 10 mg 10mg
16493957 50 mg 50mg
16473746 25 mg 25mg
16447816 100 mg 100mg
16430508 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498038 Supplier Medchem Express Supplier No. HY32718
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pelitinib (EKB-569;WAY-EKB 569) is an irreversible inhibitor of EGFR with an IC50 of 38.5 nM; also slightly inhibits Src, MEK/ERK and ErbB2 with IC50s of 282, 800, and 1255 nM, respectively.

Pelitinib (EKB-569

Specifications

Chemical Name or Material Pelitinib
CAS 257933-82-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H23ClFN5O2
Quantity 10 mg
Synonym EKB-569 WAY-EKB 569
Solubility Information DMSO : 25 mg/mL (53.43 mM; Need ultrasonic)
SMILES N#CC1=C(C2=CC(NC(/C=C/CN(C)C)=O)=C(OCC)C=C2N=C1)NC3=CC(Cl)=C(C=C3)F
Molecular Weight (g/mol) 467.92
Formula Weight 467.92
Percent Purity 96.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.