Learn More
Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections
Codice prodotto. 10392975 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Type:
Cross
Tee
Union
Diameter (English) Inner:
0.01 in.
0.02 in.
Dimensione della confezione:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Type Diameter (English) Inner Dimensione della confezione
10392975 Tee 0.02 in. Each
10422155 Union 0.02 in. Each
10149534 Cross 0.02 in. Each
10716163 Union 0.01 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10392975 Fornitore Thermo Scientific™ N. del fornitore P727
Estimated Shipment: 25-11-2026
Aggiungi al carrello
Edge
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections. The material is biocompatible for a wide range of applications.
  • Absolute zero or low dead volume formats
  • Biocompatible
TRUSTED_SUSTAINABILITY
Specifica
Type Tee
Diameter (English) Inner 0.02 in.
Description PEEK Tee;<0.57 uL Swept Volume
Diameter (Metric) Inner 12.45 mm
For Use With (Equipment) Tubing
Length (Metric) 27.43 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.