missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 360μm O.D., 10 ft
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
Specifications
| Color | Green |
| pH Range | 2 to 14 |
| Diameter (English) Inner | 125 μm (0.005 in.) |
| Diameter (Metric) Outer | 360 μm |
| Overall Length (English) | 10 ft. |
| Material | PEEK |
| Maximum Pressure | 2000 psi (138 bar) |
| Max. Operating Temperature (Metric) | 100°C |
For research use only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?