missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 360μm O.D., 10 ft

Product Code. 15761049
Change view
Click to view available options
Diameter (English) Inner:
100 μm (0.004 in.)
125 μm (0.005 in.)
150 μm (0.006 in.)
50 μm (0.002 in.)
75 μm (0.003 in.)
Color:
Black
Green
Natural
Red
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Color unitSize
15761049 50 μm (0.002 in.) Natural Each
15701059 75 μm (0.003 in.) Black Each
15741059 100 μm (0.004 in.) Red Each
15781059 125 μm (0.005 in.) Green Each
15721069 150 μm (0.006 in.) Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15761049 Supplier Trajan Scientific and Medical Supplier No. 130400200010F
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs

Specifications

Color Natural
pH Range 2 to 14
Diameter (English) Inner 50 μm (0.002 in.)
Diameter (Metric) Outer 360 μm
Overall Length (English) 10 ft.
Material PEEK
Maximum Pressure 5000 psi (345 bar)
Max. Operating Temperature (Metric) 100°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.