Learn More
Cytiva PEEK Connector Luer/M6
Product Code. 11380142 Shop All Cytiva Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Connections:
Luer Male to M6 Female
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections missing translation for 'unitSize'
11380142 Luer Male to M6 Female Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 11380142 missing translation for 'mfr' Cytiva missing translation for 'supplierNo' 11002643
In Stock
Set of five (5) polyetheretherketone Luer Lock Male/M6 Female connectors for use with the ÄKTAcrossflow tangential flow filtration system.
ÄKTAcrossflow is a fully automated tangential flow filtration system for process development and is suitable for filter screening and process optimization at small-scale as well as small-scale processing.
  • Polyetheretherketone (PEEK) connstruction
  • Connections: Luer male-M6 female
TRUSTED_SUSTAINABILITY
Specifications
Type Connector Luer
Connections Luer Male to M6 Female
Quantity Set of 5
Material PEEK
Color Black
For Use With (Equipment) ÄKTAcrossflow Tangential Flow Filtration System