Learn More
Gilac™ Pedal Container with Lid, 50 L
Product Code. 17988521
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17988521 White Each
17908591 Blue Each
17998921 Green Each
17968841 Red Each
17948451 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17988521 Supplier Gilac™ Supplier No. G614321
  • ONLINE EXCLUSIVE PRICE
    82.69 EUR
    59.90 EUR / Pack of 1
    Save 22.79 EUR
    27% Off
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Pedal Container with Lid is a compact pedal container for space saving and hands-free operation to prevent contamination.
  • Compact pedal container for space saving.
  • Hands-free operation to prevent contamination and facilitate barrier gestures, pedal prevents contamination.
  • 2 pedal mounting positions available: automatic or manual lid closing.
  • The lid (in manual lid version) can be closed by pressing the left part of the pedal.
  • Longevity and durability.
  • Perfect stability.
  • Easy to clean.
  • Colored lids for selective waste sorting.
  • Self-assembled kit: lid, body, screws and pedal delivered in the same box.
Specifications
Capacity (Metric) 50 L
Capacity (English) 13.2 gal.
Color White
Autoclavable Not Autoclavable
Material Polypropylene
Product Type Pedal Container with Lid