missing translation for 'onlineSavingsMsg'
Learn More

Pectin, MP Biomedicals™

Product Code. 11498210 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11498210

Brand: MP Biomedicals 0210258791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9000-69-5
Molecular Formula (C7H10O7)n
Synonym Poly-D-galacturonic acid methyl ester

Specifications

Color Beige to White
Physical Form Powder
Quantity 1 kg
Formula Weight 20,000 to 400,000
Chemical Name or Material Pectin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.