Learn More
Kartell™ Automatic Burette Washers
Product Code. 10395821
Change view
Click to view available options
Diameter (Metric):
170mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10395821 170mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10395821 Supplier Kartell™ Supplier No. 0021500
  • ONLINE EXCLUSIVE PRICE
    181.00 EUR
    176.91 EUR / Pack of 1
    Save 4.09 EUR
    2% Off
In Stock
Washes Mohr burettes quickly and safely. Kartell™ Automatic Burette Washers syphon contaminated waters away from burettes before refilling automatically.
Washes Mohr burettes quickly and safely. Kartell™ Automatic Burette Washers syphon contaminated waters away from burettes before refilling automatically.
  • Polyethylene rinser fills via spray nozzle situated on top of rim, above syphon point
  • Contaminated waters syphon through large diameter drain at lowest point of rinser
  • Burette baskets hold burettes and minimize breakage during process; purchased separately
  • Extra baskets can be used for pre-soaking while using rinser
  • Outlet tubing to direct contaminated waters available separately
  • Suggested basic set:
    • Burette rinser
    • 2 Burette baskets
    • 2 Extension handles for baskets
    • Burette jar
Specifications
For Use With Washing Mohr Burette
Height (Metric) 990 mm
Diameter (Metric) 170mm
For Use With (Application) Washing Mohr burettees
Product Type Automatic Burette Washer