Learn More
asecos™ PE IBC Station
Product Code. 17896526 Shop All asecos Products
Change view
Click to view available options
Capacity (English):
264.2 gal.
Unit Size:
Each
Product Code. Capacity (English) unitSize
17896526 264.2 gal. Each
1 options
Product Code. 17896526
Brand: asecos™ 24150
This item requires a price update prior to being ordered.
Request a Price for this item here.
The PE IBC Station is approved for the safe storage of water pollutants.
  • Highly resistant against oil, acid, alkali, etc.
  • Easy and safe in-house transport of the empty sump by forklift truck
Specifications
Color Blue
Capacity (English) 264.2 gal.
Capacity (Metric) 1000 L
Material Polyethylene
Type IBC Spill Pallet
Length (English) 50.39 in.
Length (Metric) 1280.00 mm
Width (English) 50.39 in.
Width (Metric) 1280.00 mm
Height (English) 34.25 in.
Height (Metric) 870.00 mm
Depth (English) 65.35 in.
Depth (Metric) 1660.00 mm
Weight (English) 187.39 lb.
Weight (Metric) 85 kg