Learn More
PD153035 Hydrochloride, MedChemExpress
Product Code. 16457667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457667 5 mg 5mg
16455696 100 mg 100mg
16434918 10 mg 10mg
16431167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16457667 Supplier Medchem Express Supplier No. HY12013
MedChemExpress PD153035 Hydrochloride (SU-5271 Hydrochloride) is a potent EGFR inhibitor with Ki and IC50 of 6 and 25 pM, respectively.
PD153035 Hydrochloride (SU-5271 Hydrochloride) is a potent EGFR inhibitor with Ki and IC50 of 6 and 25 pM, respectively.
Specifications
Chemical Name or Material PD153035 Hydrochloride
CAS 183322-45-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H15BrClN3O2
Quantity 5 mg
Synonym SU-5271 HydrochlorideAG1517 HydrochlorideZM 252868 Hydrochloride
Solubility Information DMSO : 5.125 mg/mL (12.92 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES COC1=C(C=C(C2=C1)N=CN=C2NC3=CC(Br)=CC=C3)OC.Cl
Molecular Weight (g/mol) 396.67
Formula Weight 396.67
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease