missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Antibody (rTYMP/3444), DyLight 650, Novus Biologicals™
Shop All Bio Techne ProductsDescription
PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PD-ECGF/Thymidine Phosphorylase |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rTYMP/3444 |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human PD-ECGF/Thymidine Phosphorylase protein (Uniprot: P19971 ) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?