Learn More
MedChemExpress PCI-33380
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Reference compound |
| Molecular Weight (g/mol) | 855.78 g/mol |
| Solubility | DMSO : ≥ 50 mg/mL (58.43 mM) |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| CAS | 1022899-36-0 |