Learn More
PBS, 10X Solution, Molecular Biology Grade
Product Code. 15835418 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 L
5 L
Unit Size:
1 L
100mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15835418 5 L 5 L
15815418 100 mL 100mL
15825418 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15835418 Supplier Thermo Scientific Supplier No. J75889.K8
  • ONLINE EXCLUSIVE PRICE
    184.00 EUR
    165.65 EUR / 5 L
    Save 18.35 EUR
    9% Off
 Restricted Item
In Stock
Add to basket
For biological research. Affymetrix/USB™ PBS, 10X Solution, Molecular Biology Grade is 0.2μm filtered and dispensed into durable, square nalgene bottles and upright carton.
  • Consists of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O
  • Filtered and dispensed into durable, square Nalgene bottles and upright carton with convenient dispensing spout
  • Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate
TRUSTED_SUSTAINABILITY
Specifications
pH 7.4
Physical Form Solution
Quantity 5 L
Formulation 80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O