Learn More
Pazopanib Hydrochloride, MedChemExpress
Product Code. 16495686
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495686 100 mg 100mg
16474908 10 mg 10mg
16461157 50 mg 50mg
16421626 500 mg 500mg
16421026 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16495686 Supplier Medchem Express Supplier No. HY12009
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Pazopanib Hydrochloride (GW786034 Hydrochloride) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms with an IC50 of 10, 30, 47, 84, 74, 140 and 146 nM, respectively.
Pazopanib Hydrochloride (GW786034 Hydrochloride) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms with an IC50 of 10, 30, 47, 84, 74, 140 and 146 nM, respectively.
Specifications
Chemical Name or Material Pazopanib Hydrochloride
CAS 635702-64-6
Color White
Purity Grade Notes Research
Molecular Formula C21H24ClN7O2S
Quantity 100 mg
Synonym GW786034 (Hydrochloride)
Solubility Information DMSO : 10 mg/mL (21.10 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=CC=C(C=C1S(N)(=O)=O)NC2=NC=CC(N(C3=CC4=NN(C(C)=C4C=C3)C)C)=N2.Cl
Molecular Weight (g/mol) 473.98
Formula Weight 473.98
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease