Learn More
Intelsius™ PathoSorb Absorbent Sheet
Product Code. 17318589
Encompass_Preferred
Change view
Click to view available options
Type:
Absorbent Sample Sheet
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17318589 Absorbent Sample Sheet Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17318589 Supplier Intelsius™ Supplier No. AB007
Estimated Shipment: 24-11-2026
The PathoSorb Absorbent Sheet is a lint free absorbent sheet that reduces the contamination risk and satisfies category A and B transport regulations when transporting samples.
  • 600 mL saline absorbency capacity
  • 693 mL water absorbency capacity
  • Lint free
  • Super Absorbent
  • Suitable in supporting UN3373 Category B and UN2900/UN2814 Category A transport regulations 
Specifications
Type Absorbent Sample Sheet
Dimensions (L x W) 470 x 295 mm (18.5 x 11.6 in.)
Capacity 600 mL