missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress XBP1 Nuclear Translocation Assay
Description
The PathHunter™ eXpress XBP1 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the IRE1/XBP1 pathway. This kit assesses ligand (e.g. thapsigargin) based pathway activation via detection of resulting XBP1 nuclear translocation. This product uses EFC technology. The included cells overexpress PL-tagged XBP1 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of XBP1 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Translocation |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Thapsigargin |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | XBP1 |
| Cell Line | U2OS |
| Target | XBP1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?